Learn More
Essendant GLASSES10/CTBE
Supplier:  Essendant MMMSF3701XSGAB
Encompass_Preferred
3M Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes eliminating the need to stock multiple frame sizes at worksites Secure snug fit keeps eyewear in place without slippage even when the wearer is in motion Anti-fog coating helps keep lenses free of moisture in hot and humid conditions Polycarbonate lenses absorb 99 9pct UVA and UVB up to 380 nm SecureFit 3700 OTG Series Designed to offer eye protection with all-day wearability for workers with prescription eyeglasses Safety Glasses Style- Over-the-Glasses Frame Color- Blue Frame Material- Plastic Lens Material- Polycarbonate
Catalog No. 50-000-35973