Learn More
Essendant GLASSESEQUALZR BE/BE MIR
Supplier:  Essendant ANS21301
Encompass_Preferred
Stylish safety glasses have a modern cutting edge design Soft rubber nose pads for superior comfort and fit Dual lenses with an 8-base curve offer wraparound protection Sport-grip temples eliminate slippage and help glasses sit comfortably High performance polycarbonate lenses provide 99 9pct UV protection Safety eyewear meets ANSI Z87 1plus impact standard Style- Wraparound Frame Color- Blue Lens Material- Polycarbonate Lens Tint- Blue Mirror
Catalog No. 50-000-20457