Learn More
Essendant Law Over the Glasses Safety Glasses, Clear Anti-Fog Lens
Catalog No. 2790921
Encompass_Preferred
Breaks through the traditional look of over-the-glasses protective eyewear. Durable polycarbonate frame designed to be worn over prescription glasses. Five-position ratcheting temple design for optimum fit. Nonslip temple inserts provide comfort and grip. Anti-fog, scratch-resistant and filters 99.9% of UV rays.
Catalog No. 27-909-21 Supplier Essendant Supplier No. CRWOG110AF
Breaks through the traditional look of over-the-glasses protective eyewear. Durable polycarbonate frame designed to be worn over prescription glasses. Five-position ratcheting temple design for optimum fit. Nonslip temple inserts provide comfort and grip. Anti-fog, scratch-resistant and filters 99.9% of UV rays.