Learn More
Essendant GLASSESSFTYFOAM12BK
Supplier:  Essendant ANS65335
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees With KleenGuard Nemesis Foam Safety Glasses you get sleek modern eyewear with durable lightweight construction That is just one of the reasons why millions of people wear them for protection both at work and at home KleenGuard Nemesis Foam Safety Glasses have polycarbonate lenses that provide 99 9pct UVA/ UVB/UVC protection to help prevent eye damage like cataracts retinal damage and other conditions that can cause temporary vision loss The wraparound frame has a base curve of 8 that extends the range of peripheral protection as well as soft touch temples for comfort KleenGuard Nemesis Foam Safety Glasses have a patented nose piece designed to be extremely flexible to channel sweat away from the eyes and to help prevent slippage down the nose all for long-term comfort Each pair of KleenGuard Nemesis Foam Safety Glasses come w
Catalog No. 50-000-20669