Learn More
Electron Microscopy Sciences Glassine Negative Envelope Plain, 3-1/4 x 4"
SDP
Supplier:  Electron Microscopy Sciences 74054
Encompass_Preferred
Glassine envelopes, acid free for negative storage. Open end, thumb-cut, side-seamed.
Use: 3-1/4 x 4"
Catalog No. 50-261-68