Learn More
Electron Microscopy Sciences Glassine Negative Envelope Plain, 6.5 x 9 cm
SDP
Catalog No. 5026156
Encompass_Preferred
Glassine envelopes, acid free for negative storage. Open end, thumb-cut, side-seamed.
Use: 6.5 x 9 cm
Catalog No. 50-261-56 Supplier Electron Microscopy Sciences Supplier No. 74080
Glassine envelopes, acid free for negative storage. Open end, thumb-cut, side-seamed.
Use: 6.5 x 9 cm