Learn More
Andwin Scientific GLASSINE WEIGH PAPER 6X6 500PK
SDP
Catalog No. 501305926
Encompass_Preferred
The 6x6 small glassine weighing paper is moisture-resistant and non-absorbent to allow complete sample transfer and keep balance pans clean. It is sold in a pack of 500.
Catalog No. 50-130-5926 Supplier Andwin Scientific Supplier No. 133804A
The 6x6 small glassine weighing paper is moisture-resistant and non-absorbent to allow complete sample transfer and keep balance pans clean. It is sold in a pack of 500.