Learn More
Electron Microscopy Sciences Glassine Weighing Dish
SDP
Supplier:  Electron Microscopy Sciences 7008401
Encompass_Preferred
Weighing samples is made easier with these newly introduced Glassine Weighing Boats. No static build-up because Glassine is free of static electricity that plagues disposable plastic weighing dishes. Contaminant free because pure glassine is an ultra-clean paper product. Preformed 3.5 diameter dish. Chocolate colored glassine reveals every particle of material with color contrast.
Catalog No. 50-334-76