Learn More
MSC PortionPac Chemical Glass Cleaner, Streak-Free, Biodegradable, VOC-Free, Green Seal GS-37
Catalog No. 502835650
Encompass
PortionPac Chemical Corporation's GlassPac Glass Cleaner is a streak-free, biodegradable, VOC-free, and fragrance-free cleaning solution. It is formulated to effectively clean various surfaces including glass, mirrors, stainless steel, copper, brass, Formica, vinyl, Plexiglas, and Lexan, and meets Green Seal™ Standard GS-37.
  • Provides streak-free cleaning
  • Features a biodegradable formulation
  • VOC free formulation
  • Fragrance free
  • Meets Green Seal™ Standard GS-37
  • Concentrated volume
  • Minimized and recycled packaging
  • Offers protective limits on VOCs, human, and environmental toxicity
  • Effective for cleaning various surfaces
  • Environmentally conscious formulation
  • Safe for use with no defined hazards by OSHA
Catalog No. 50-283-5650 Supplier MSC Supplier No. 38688859
PortionPac Chemical Corporation's GlassPac Glass Cleaner is a streak-free, biodegradable, VOC-free, and fragrance-free cleaning solution. It is formulated to effectively clean various surfaces including glass, mirrors, stainless steel, copper, brass, Formica, vinyl, Plexiglas, and Lexan, and meets Green Seal™ Standard GS-37.
  • Provides streak-free cleaning
  • Features a biodegradable formulation
  • VOC free formulation
  • Fragrance free
  • Meets Green Seal™ Standard GS-37
  • Concentrated volume
  • Minimized and recycled packaging
  • Offers protective limits on VOCs, human, and environmental toxicity
  • Effective for cleaning various surfaces
  • Environmentally conscious formulation
  • Safe for use with no defined hazards by OSHA