Learn More
Perkin Elmer US LLC Glass-Ring for DPC Caps of Standard 75 mL Vessels
Catalog No. 502111685
Encompass_Preferred
DPC Glass-Ring for Digestion Vessel caps with pressure control for Standard 75 mL (40 Bar) Digestion Vessel.
Catalog No. 50-211-1685 Supplier Perkin Elmer US LLC Supplier No. N3132003
DPC Glass-Ring for Digestion Vessel caps with pressure control for Standard 75 mL (40 Bar) Digestion Vessel.