Learn More
MSC Betco Glass Cleaner, Concentrated, Liquid, 2 L, 4 Bottles
Catalog No. 502868581
Encompass
This Betco concentrated glass cleaner is supplied in 2 L plastic bottles and is formulated for effective cleaning of glass surfaces.
  • Liquid form
  • Bottle container type
  • Container size: 2 L
  • Pleasant scent
  • Concentrated formula
  • Designed for glass surfaces
  • EcoLogo certified
  • Green Seal certified
  • USDA Certified BioBased
  • Made from recycled content
Catalog No. 50-286-8581 Supplier MSC Supplier No. 35174168
This Betco concentrated glass cleaner is supplied in 2 L plastic bottles and is formulated for effective cleaning of glass surfaces.
  • Liquid form
  • Bottle container type
  • Container size: 2 L
  • Pleasant scent
  • Concentrated formula
  • Designed for glass surfaces
  • EcoLogo certified
  • Green Seal certified
  • USDA Certified BioBased
  • Made from recycled content