Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10
GSA_VA
Catalog No. 22749754
Change view
Click to view available options
Quantity:
100 Pack
50 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-749754 50 Pack
VT87146 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-749754 Supplier Kova Plastics by Alltrista Supplier No. 87157E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis.
  • Made from optically clear plastic
  • Glasstic Slide features 10 individual fully segregated specimen wells
  • Capillary action reduces the likelihood of pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
  • Correlate well with results obtained by using a hemocytometer: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Slide 10
For Use With (Application) Microscopy
Gridded No
Material Plastic with the optical qualities of glass
No. of Wells 10
Quantity 50 Pack