Learn More
Kova Plastics by Alltrista Glasstic™ Slide II
Catalog No. 22741207
Change view
Click to view available options
Gridded:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gridded
22-741207 Yes
22-741215 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-741207 Supplier Kova Plastics by Alltrista Supplier No. 87100E
Add to Cart
Add to Cart
Four-chambered slide
Slide ll is great for accurate, reproducible, and efficient standardized microscopic analysis.
  • Made from optically clear plastic
  • Each Slide ll features four individual fully segregated specimen wells
  • Available with or without integrated counting grids
  • Slides that include integrated counting grids provide results that have high correlation with results obtained with precision hemocytometers
  • Capillary action reduces the likelihood of pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
Specifications
Type Slide II
For Use With (Application) Microscopy
Gridded Yes
Material Plastic with the optical qualities of glass
No. of Wells 4
Quantity 100