Learn More
Kova Plastics by Alltrista Glasstic™ Slide II
GSA_VA
Catalog No. 22741215
Change view
Click to view available options
Gridded:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gridded
22-741215 No
22-741207 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-741215 Supplier Kova Plastics by Alltrista Supplier No. 87118E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Four-chambered slide
Slide ll is great for accurate, reproducible, and efficient standardized microscopic analysis.
  • Made from optically clear plastic
  • Each Slide ll features four individual fully segregated specimen wells
  • Available with or without integrated counting grids
  • Slides that include integrated counting grids provide results that have high correlation with results obtained with precision hemocytometers
  • Capillary action reduces the likelihood of pipetting errors
  • Suspends sediments homogeneously throughout the specimen
  • Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
  • Reproducible fluid distribution throughout each chamber
  • Chamber walls prevent specimen leakage and cross-contamination
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Slide II
For Use With (Application) Microscopy
Gridded No
Material Plastic with the optical qualities of glass
No. of Wells 4
Quantity 100