Learn More
Research Products International Corp Low Background Glass Scintillation Vials, 20ml Capacity, 22mm Poly Lined Plastic Unattached Screw Caps, 500 per Case
SDP
Catalog No. 50212999
Encompass
Low background screw thread vial manufactured from borosilicate glass to our own exact vial and stringent specifications. Special attention given to all critical dimensions such as wall thickness, vial diameter and length.
Catalog No. 50-212-999 Supplier Research Products International Corp Supplier No. 121053
Low background screw thread vial manufactured from borosilicate glass to our own exact vial and stringent specifications. Special attention given to all critical dimensions such as wall thickness, vial diameter and length.