Learn More
Research Products International Corp Glassware Ring Anchors, Size 1, Fits Flasks 125 -500ml, Weight: 1/2 Pound (227 Grams)
SDP
Catalog No. 50212928
Encompass
Glassware anchors are plastic coated lead weights offered in a one-piece ring shape for use with flasks. Each anchor has a durable and chemically resistant vinyl coating that is non-contaminating. Bright red color helps identify and increase visibility of carefully placed flasks.
WARNING  This product can expose you to chemicals including Lead, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-212-928 Supplier Research Products International Corp Supplier No. CDS0523
Glassware anchors are plastic coated lead weights offered in a one-piece ring shape for use with flasks. Each anchor has a durable and chemically resistant vinyl coating that is non-contaminating. Bright red color helps identify and increase visibility of carefully placed flasks.
WARNING  This product can expose you to chemicals including Lead, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.