Learn More
MESA LABS BOZEMAN MFG FACILITY Mesa Labs PAK-G Glassware Kit, Sterilized Components, Assay Tubes and Pipettes Included, 41 Units
SDP
Catalog No. NC0869510
Encompass
The Mesa Labs PAK-G Glassware Kit provides essential sterile glassware components designed for accurate population assays of biological indicators. This comprehensive kit includes tubes, pipettes, and glass beads, all packaged securely in a corrugated box to ensure sterility and integrity for laboratory use. Ideal for performing dilutions in biological assays, it differentiates itself with its specifically designed components optimal for precise laboratory applications.
Catalog No. NC0869510 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. PAKG
The Mesa Labs PAK-G Glassware Kit provides essential sterile glassware components designed for accurate population assays of biological indicators. This comprehensive kit includes tubes, pipettes, and glass beads, all packaged securely in a corrugated box to ensure sterility and integrity for laboratory use. Ideal for performing dilutions in biological assays, it differentiates itself with its specifically designed components optimal for precise laboratory applications.