Learn More
SP Hotpack™ Glassware Washer Accessory, Watch Glass Insert
Catalog No. 15365024 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds:
Up to 40 pieces <125 mm dia.
Catalog No. Holds
15-365-024 Up to 40 pieces <125 mm dia.
1 options
Catalog No. 15-365-024 Supplier SP Hotpack™ Supplier No. 100519231
Add to Cart
Add to Cart
Accessory designed for use with SP Hotpack™ undercounter, freestanding, and vertical stacking glassware washers.
  • Accessories maximize space, improve efficiency, and protect specialty glassware
  • For use in standard open racks
  • Made of stainless steel for long life and durability
Specifications
Material Stainless Steel
Product Type Insert
Holds Up to 40 pieces <125 mm dia.
For Use With (Equipment) Watch Glass