Description
Spindle racks are available in multiple sizes and configurations to best fit your application:
- Top and bottom spindle rack with full spindle on both bottom and top.
- Top and bottom spindle rack with full spindle on bottom and half spindle/half standard on top.
- Top and bottom spindle rack with full spindle on bottom and half size spindle on top to accommodate tall glass on bottom rack.
- Bottom spindle rack only with half spindle and half standard.
- Bottom spindle rack only with full spindle bottom.
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Rack |
| Rack Type | Top and Bottom Spindle Rack, Full Spindle Bottom and Top |
| For Use With (Equipment) | SP Hotpack Spindle Ready Glassware Washers |
Used with SP Hotpack™ Spindle Rack Ready Washers Only