Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glassy Carbon Lid;Top Dia (mm), typically 50;Bottom Dia (mm), 36, Thermo Scientific™

Catalog No. AA39021KT
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Shrink-wrapped
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA39021KT 1 Ea. Shrink-wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39021KT Supplier Thermo Scientific Chemicals Supplier No. 39021KT
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Shrink-wrapped
Quantity 1 Ea.
Chemical Name or Material Glassy Carbon Lid;Top Dia (mm)
TSCA U
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.