Learn More
Medchemexpress LLC Glatiramer acetate | 147245-92-9 | MFCD04113403 | 98.0% | 623.7 g/mol | C25H45N5O13 | 5 MG
SDP
Catalog No. 5000263460
Encompass_Preferred
Glatiramer acetate is a synthetic polypeptide used as an immunomodulator in multiple sclerosis research. It is supplied as a solid powder for laboratory research applications and is labeled for research use only; it is not for patient or clinical use.
  • Synthetic copolymer of four amino acids suitable for immunological studies.
  • Used as an immune modulator in multiple sclerosis models and related research.
  • Available in small milligram pack sizes for controlled experimental dosing.
  • High reported purity supports reproducible experimental results.
  • Provided as a solid powder for flexible preparation and dosing in vitro or in vivo.
Catalog No. 50-002-63460 Supplier Medchemexpress LLC Supplier No. HY1095205MG
Glatiramer acetate is a synthetic polypeptide used as an immunomodulator in multiple sclerosis research. It is supplied as a solid powder for laboratory research applications and is labeled for research use only; it is not for patient or clinical use.
  • Synthetic copolymer of four amino acids suitable for immunological studies.
  • Used as an immune modulator in multiple sclerosis models and related research.
  • Available in small milligram pack sizes for controlled experimental dosing.
  • High reported purity supports reproducible experimental results.
  • Provided as a solid powder for flexible preparation and dosing in vitro or in vivo.