Learn More
Apexbio Technology LLC Glatiramer acetate 147245-92-9 5mg
SDP
Catalog No. 5000063901
Encompass_Preferred
Glatiramer acetate (GA) is a synthetic immunomodulatory polypeptide structurally composed of random sequences of four amino acids GA modulates immune responses mainly through affecting dendritic cell (DC) function In vitro studies indicate GA suppresses IL-12p70 secretion in immature DC following lipopolysaccharide stimulation promoting DC differentiation favoring Th2 lineage cells and elevated IL-10 production Further in vivo studies reveal GA treatment alters cytokine profiles and enhances expression of neurotrophic factors such as BDNF NT-3 and NT-4 within specific brain regions of experimental autoimmune encephalomyelitis mouse models highlighting its utility in immune research and autoimmune disease investigations
Catalog No. 50-000-63901 Supplier Apexbio Technology LLC Supplier No. B49785
Glatiramer acetate (GA) is a synthetic immunomodulatory polypeptide structurally composed of random sequences of four amino acids GA modulates immune responses mainly through affecting dendritic cell (DC) function In vitro studies indicate GA suppresses IL-12p70 secretion in immature DC following lipopolysaccharide stimulation promoting DC differentiation favoring Th2 lineage cells and elevated IL-10 production Further in vivo studies reveal GA treatment alters cytokine profiles and enhances expression of neurotrophic factors such as BDNF NT-3 and NT-4 within specific brain regions of experimental autoimmune encephalomyelitis mouse models highlighting its utility in immune research and autoimmune disease investigations