Learn More
Medchemexpress LLC HY-N2113 5mg Medchemexpress, Glaucocalyxin B CAS:80508-81-2 Purity:>98%
SDP
Catalog No. 501933886
Encompass_Preferred
Medchemexpress, HY-N2113 5mg Glaucocalyxin B CAS:80508-81-2 Glaucocalyxin B is an ent kaurane diterpenoid isolated from the Chinese traditional medicine Rabdosia japonica with anticancer and antitumor activity; decreases the growth of HL-60 cells with an IC 50 of approximately 5.86 μM at 24 h. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3886 Supplier Medchemexpress LLC Supplier No. HYN21135MG
Medchemexpress, HY-N2113 5mg Glaucocalyxin B CAS:80508-81-2 Glaucocalyxin B is an ent kaurane diterpenoid isolated from the Chinese traditional medicine Rabdosia japonica with anticancer and antitumor activity; decreases the growth of HL-60 cells with an IC 50 of approximately 5.86 μM at 24 h. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.