Learn More
Aviva Systems Biology Corp GLB1 Antibody - N-terminal region (ARP61554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLB1. It was validated on Western Blot. . Alternate names: EBP, ELNR1, MPS4BApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-287-407