Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GLB1/Beta-galactosidase Antibody (YA2816) | 100 UL
SDP

Catalog No. 5000444557
Encompass_Preferred

GLB1/Beta-galactosidase Antibody (YA2816) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting GLB1/Beta-galactosidase. It is suitable for applications such as Western Blot (WB) and Flow Cytometry (FC). This antibody is affinity purified and supplied in a buffered saline solution containing BSA, sodium azide, and glycerol, ensuring stability for up to 1 year when stored at -20°C.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets GLB1/Beta-galactosidase
  • Reactivity in human, mouse, and rat
  • Predicted molecular weight of 76 kDa; observed molecular weight of 95 kDa
  • Affinity purified
  • Applications include WB (1:500-1:1000) and FC (1:50-1:100)
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles

Catalog No. 50-004-44557 Supplier Medchemexpress LLC Supplier No. HYP83071100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GLB1/Beta-galactosidase Antibody (YA2816) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting GLB1/Beta-galactosidase. It is suitable for applications such as Western Blot (WB) and Flow Cytometry (FC). This antibody is affinity purified and supplied in a buffered saline solution containing BSA, sodium azide, and glycerol, ensuring stability for up to 1 year when stored at -20°C.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets GLB1/Beta-galactosidase
  • Reactivity in human, mouse, and rat
  • Predicted molecular weight of 76 kDa; observed molecular weight of 95 kDa
  • Affinity purified
  • Applications include WB (1:500-1:1000) and FC (1:50-1:100)
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.