Learn More
Medchemexpress LLC GlcCer (d18:1/18:0) | 95119-86-1 | 99.93% | 728.09 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1700355MG
Encompass_Preferred
GlcCer (d18:1/18:0) (C18 Glucosyl (β) ceramide (d18:1/18:0)) is a glycosphingolipid. It is promising for research of Parkinson disease and dementia with Lewy bodies.
  • A glycosphingolipid
  • Promising for research of Parkinson disease and dementia with Lewy bodies
  • Powder: -20°C for 3 years
  • In solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-91297