Learn More
Aviva Systems Biology Corp GLCCI1 Antibody - middle region (ARP52598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLCCI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GLCCI1. Alternate names: FAM117C, GIG18, GCTR, TSSN1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-738