Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GlcNAc beta(1-3)GalNAc-alpha-Thr 97.0+%, TCI America™
SDP

Catalog No. G03412MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03412MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03412MG Supplier TCI America Supplier No. G03412MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 286959-52-2
Molecular Formula C20H35N3O13
Molecular Weight (g/mol) 525.508
InChI Key BMSPZGKRTLZXAV-RGVQASSVSA-N
Synonym O-[2-Acetamido-2-deoxy-3-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-alpha-D-galactopyranosyl]-L-threonine
PubChem CID 53384351
IUPAC Name (2S,3R)-3-[(2S,3S,4R,5R,6S)-3-acetamido-4-[(2R,3S,4R,5S,6S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-aminobutanoic acid
SMILES CC(C(C(=O)O)N)OC1C(C(C(C(O1)CO)O)OC2C(C(C(C(O2)CO)O)O)NC(=O)C)NC(=O)C

Specifications

Color White
Quantity 2 mg
Formula Weight 525.51
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-3)GalNAc-alpha-Thr
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.