Learn More
MOLTOX Molecular Toxicology Inc GLDN SRN HAMPSTER LIVER S9,ML
SDP
Catalog No. NC1955615
Encompass
golden syrian hampster liver s9, male, pb-bnf inducedin 0.15 M KCl 5ml per vial. *Non Returnable*
Catalog No. NC1955615 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 15205.5
Add to Cart
Add to Cart
golden syrian hampster liver s9, male, pb-bnf inducedin 0.15 M KCl 5ml per vial. *Non Returnable*