Learn More
TARGETMOL CHEMICALS INC GLECAPREVIR 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T512625MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Glecaprevir (ABT-493) is an HCV NS3/4A protease inhibitor (IC50s 3.5-11.3 nM). purity: 99%
Catalog No. 50-257-9808