Learn More
Medchemexpress LLC HY-17634 5mg , Glecaprevir CAS:1365970-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY176345MG
Encompass_Preferred
HY-17634 5mg Glecaprevir CAS: 1365970-03-1 Glecaprevir is a novel HCV NS3/4A protease inhibitor, with IC50 values ranging from 3.5 to 11.3 nM. Glecaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 4.09 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9778