Learn More
Glen Research Glen-Pak™ DNA Purification Cartridge, 96/pk
Catalog No. NC1731884
Encompass_Preferred
Glen-Pak™ DNA and RNA cartridges have advantages over Poly-Pak cartridges in that a single loading of the diluted crude deprotection solution is all that is necessary. Also, the range of purification has been extended to 100+ using DMT-on oligos. Glen-Pak cartridges have similar performance to Fluoro-Pak cartridges but without the need for the fluorous DMT group at the 5' terminus and special phosphoramidites, so the cost is lower. In addition, Glen-Pak cartridges allow purification of virtually the complete range of dyes and modifiers.
Catalog No. NC1731884 Supplier Glen Research Supplier No. 60510096
Glen-Pak™ DNA and RNA cartridges have advantages over Poly-Pak cartridges in that a single loading of the diluted crude deprotection solution is all that is necessary. Also, the range of purification has been extended to 100+ using DMT-on oligos. Glen-Pak cartridges have similar performance to Fluoro-Pak cartridges but without the need for the fluorous DMT group at the 5' terminus and special phosphoramidites, so the cost is lower. In addition, Glen-Pak cartridges allow purification of virtually the complete range of dyes and modifiers.