Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glen Research Glen Research Spacer Phosphoramidite, Spacer 9, 0.25 g

Catalog No. NC3983918
Encompass_Preferred

This spacer phosphoramidite is used for inserting a spacer arm into an oligonucleotide. Multiple additions of this compound can be made when a longer spacer is needed.

  • Chemical name: 9-O-Dimethoxytrityl-triethylene glycol,1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite.
  • Formula: C36H49N2O7P.
  • M.W.: 652.77.
  • F.W.: 212.14.
  • CAS number: 146668-73-7.
  • Diluent: Anhydrous acetonitrile.
  • Recommended storage: Freezer storage, –10°C to –30°C, dry.
  • Stability in solution: 2–3 days.
  • Pack size: 0.25 g.

Catalog No. NC3983918 Supplier Glen Research Supplier No. 10190902E
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This spacer phosphoramidite is used for inserting a spacer arm into an oligonucleotide. Multiple additions of this compound can be made when a longer spacer is needed.

  • Chemical name: 9-O-Dimethoxytrityl-triethylene glycol,1-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite.
  • Formula: C36H49N2O7P.
  • M.W.: 652.77.
  • F.W.: 212.14.
  • CAS number: 146668-73-7.
  • Diluent: Anhydrous acetonitrile.
  • Recommended storage: Freezer storage, –10°C to –30°C, dry.
  • Stability in solution: 2–3 days.
  • Pack size: 0.25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.