Learn More
Electron Microscopy Sciences Glendur Cloth PSA, N/H, 8" diameter
SDP
Catalog No. 50997226
Encompass_Preferred
A fine stainless steel mesh on an aluminum foil polyethylene backing. Extremely durable material and, excellent removable rate when used with a diamond compound in the 45 to 60 micron range. Recommended for coarse and intermediate polishing. Available with Pressure Sensitive Adhesive (PSA) without hole.
Type: no hole
Diameter: 8"
Catalog No. 50-997-226 Supplier Electron Microscopy Sciences Supplier No. 5033205
A fine stainless steel mesh on an aluminum foil polyethylene backing. Extremely durable material and, excellent removable rate when used with a diamond compound in the 45 to 60 micron range. Recommended for coarse and intermediate polishing. Available with Pressure Sensitive Adhesive (PSA) without hole.
Type: no hole
Diameter: 8"