Learn More
Electron Microscopy Sciences Lapping Vehicle Formula C Lube 16 OZ
SDP
Catalog No. 50997378
Encompass_Preferred
For oil soluble compounds, such as Glennel UC and US, specially designed to enhance cutting action and leave a superior finish.
Catalog No. 50-997-378 Supplier Electron Microscopy Sciences Supplier No. 5037605
For oil soluble compounds, such as Glennel UC and US, specially designed to enhance cutting action and leave a superior finish.