Learn More
Electron Microscopy Sciences Lapping Vehicle Glenoil Lube 16 OZ
SDP
Catalog No. 50997377
Encompass_Preferred
Lapping vehicle is ideal l for all Glennel UB Compounds and Diamond Sprays. It assures continued separation of diamond particles and constant removal of lap debris.
Catalog No. 50-997-377 Supplier Electron Microscopy Sciences Supplier No. 5037601
Lapping vehicle is ideal l for all Glennel UB Compounds and Diamond Sprays. It assures continued separation of diamond particles and constant removal of lap debris.