Learn More
Medchemexpress LLC Glepaglutide acetate | 99.5% | 4376.06 | 25 MG
SDP
Catalog No. 5000426929
Encompass_Preferred
Glepaglutide acetate is a long-acting GLP-2 analogue and a potent GLP-2R agonist. It reduces faecal output and increases intestinal absorption. Glepaglutide acetate alleviates small intestinal inflammation.
  • Can be used in research of inflammatory bowel disease (IBD)
  • Can be used in research of Crohn's disease
  • Reduces faecal output
  • Increases intestinal absorption
  • Alleviates small intestinal inflammation
Catalog No. 50-004-26929 Supplier Medchemexpress LLC Supplier No. 25MGHYP2221B
Glepaglutide acetate is a long-acting GLP-2 analogue and a potent GLP-2R agonist. It reduces faecal output and increases intestinal absorption. Glepaglutide acetate alleviates small intestinal inflammation.
  • Can be used in research of inflammatory bowel disease (IBD)
  • Can be used in research of Crohn's disease
  • Reduces faecal output
  • Increases intestinal absorption
  • Alleviates small intestinal inflammation