Learn More
Medchemexpress LLC Glepaglutide acetate | 99.5% | 4376.06 | 5 MG
SDP
Catalog No. 5000423121
Encompass_Preferred
Glepaglutide acetate is a long-acting GLP-2 analogue and a potent GLP-2R agonist. It functions by reducing faecal output and increasing intestinal absorption. This compound also helps in alleviating small intestinal inflammation, making it suitable for research applications in inflammatory bowel disease (IBD) and Crohn's disease.
  • Long-acting GLP-2 analogue
  • Potent GLP-2R agonist
  • Reduces faecal output
  • Increases intestinal absorption
  • Alleviates small intestinal inflammation
  • Suitable for inflammatory bowel disease (IBD) research
  • Suitable for Crohn's disease research
Catalog No. 50-004-23121 Supplier Medchemexpress LLC Supplier No. HYP2221B5MG
Glepaglutide acetate is a long-acting GLP-2 analogue and a potent GLP-2R agonist. It functions by reducing faecal output and increasing intestinal absorption. This compound also helps in alleviating small intestinal inflammation, making it suitable for research applications in inflammatory bowel disease (IBD) and Crohn's disease.
  • Long-acting GLP-2 analogue
  • Potent GLP-2R agonist
  • Reduces faecal output
  • Increases intestinal absorption
  • Alleviates small intestinal inflammation
  • Suitable for inflammatory bowel disease (IBD) research
  • Suitable for Crohn's disease research