Learn More
Medchemexpress LLC Glesatinib hydrochloride | 2319837-80-2 | 98.0% | 656.17 | 1 ML
SDP
Catalog No. 5000402795
Encompass_Preferred
Glesatinib hydrochloride is an orally active, potent dual inhibitor of MET and SMO. This tyrosine kinase inhibitor also antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).
  • Orally active and potent MET/SMO dual inhibitor.
  • Tyrosine kinase inhibitor.
  • Antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).
Catalog No. 50-004-02795 Supplier Medchemexpress LLC Supplier No. HY19642A10MM/1ML
Glesatinib hydrochloride is an orally active, potent dual inhibitor of MET and SMO. This tyrosine kinase inhibitor also antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).
  • Orally active and potent MET/SMO dual inhibitor.
  • Tyrosine kinase inhibitor.
  • Antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC).