Learn More
Medchemexpress LLC Glesatinib hydrochloride | 2319837-80-2 | 98.01% | 656.17 | 100 MG
SDP
Catalog No. 5000401518
Encompass_Preferred
Glesatinib hydrochloride (MGCD265 hydrochloride) is an orally active, potent MET/SMO dual inhibitor. This tyrosine kinase inhibitor also antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC). It is intended for research use only and not for patient use.
  • Orally active
  • Potent MET/SMO dual inhibitor
  • Tyrosine kinase inhibitor
  • Antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC)
Catalog No. 50-004-01518 Supplier Medchemexpress LLC Supplier No. HY19642A100MG
Glesatinib hydrochloride (MGCD265 hydrochloride) is an orally active, potent MET/SMO dual inhibitor. This tyrosine kinase inhibitor also antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC). It is intended for research use only and not for patient use.
  • Orally active
  • Potent MET/SMO dual inhibitor
  • Tyrosine kinase inhibitor
  • Antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC)