Learn More
Medchemexpress LLC HY-19642A 5mg Medchemexpress, Glesatinib (hydrochloride) CAS:1123838-51-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19642A5MG
Encompass_Preferred
Medchemexpress, HY-19642A 5mg Glesatinib (hydrochloride) CAS:1123838-51-6 Glesatinib hydrochloride (MGCD265 hydrochloride) is an orally active, potent MET/SMO dual inhibitor. Glesatinib hydrochloride, a tyrosine kinase inhibitor, antagonizes P-glycoprotein (P-gp) mediated multidrug resistance (MDR) in non-small cell lung cancer (NSCLC)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0868