Learn More
AUTOMATION TRAINER GLH QC Kit - Aqueous Reagents, 384-Well Verification Microplates, VRP, Mini-Reader
SDP
Catalog No. NC2293250
Encompass_Preferred
Quality Simplified. The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution.Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.- Characterizes error limits of volume transfers (1 nL to 10 mL)- Compatible with any manual or automated liquid handling instrument (up to 1536-channels)- Test Serial Dilutions (up to 48 targets in a single plate)- Diluent-Dispense-Insensitive with GLH Verification Microplates- No manual pipetting required with 96-Well and 384-Well GLH Verification Microplates- Correct any reader for NIST-Traceable results with the GLH Validation Reference PlateLegal Disclaimer: For research use only, not for diagnostic procedures.
This item is not returnable. View return policy
Catalog No. NC2293250 Supplier AUTOMATION TRAINER Supplier No. ATQCKITAQ384
Please contact your sales representative for a quote or further ordering instructions.
Quality Simplified. The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution.Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.- Characterizes error limits of volume transfers (1 nL to 10 mL)- Compatible with any manual or automated liquid handling instrument (up to 1536-channels)- Test Serial Dilutions (up to 48 targets in a single plate)- Diluent-Dispense-Insensitive with GLH Verification Microplates- No manual pipetting required with 96-Well and 384-Well GLH Verification Microplates- Correct any reader for NIST-Traceable results with the GLH Validation Reference PlateLegal Disclaimer: For research use only, not for diagnostic procedures.