Learn More
AUTOMATION TRAINER GLH 96 READER REAGENT
SDP
Catalog No. NC3727935
Encompass_Preferred
Universal Validation Reference Plate (VRP) verifies microplate reader performance and auto-generates reference or standard curves to validate GLH QC Kit test results to achieve NIST Traceability.- Test Photometric Accuracy and Linearity- Average Internal Error: 0.5%- In-Field NIST Traceability of Reader- Maintain Chain of Custody- Excel-based software for easy validation- NIST-Traceable Standards- Just-In-Time Manufactured- Works with most readers- Auto-generate reference or standard curves for volumetric verification and calibration- Supports use of verification microplates for improved test performanceGLH-Tracker Desktop software included with purchase of Universal Validation Reference Plate (VRP).The VRP can be used in conjunction with the 96-Well and 384-Well GLH Verification Microplates to improve test performance.
Catalog No. NC3727935 Supplier AUTOMATION TRAINER Supplier No. ATQCRA96R
Add to Cart
Add to Cart
Request a Quote
Universal Validation Reference Plate (VRP) verifies microplate reader performance and auto-generates reference or standard curves to validate GLH QC Kit test results to achieve NIST Traceability.- Test Photometric Accuracy and Linearity- Average Internal Error: 0.5%- In-Field NIST Traceability of Reader- Maintain Chain of Custody- Excel-based software for easy validation- NIST-Traceable Standards- Just-In-Time Manufactured- Works with most readers- Auto-generate reference or standard curves for volumetric verification and calibration- Supports use of verification microplates for improved test performanceGLH-Tracker Desktop software included with purchase of Universal Validation Reference Plate (VRP).The VRP can be used in conjunction with the 96-Well and 384-Well GLH Verification Microplates to improve test performance.