Learn More
AUTOMATION TRAINER GLH QC Kit Lite - Aqueous Reagents Set and 384-Well Verification Microplates
SDP
Catalog No. NC2293256
Encompass_Preferred
The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution. Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.Characterizes accuracy and precision of volume transfersCan be used for hardware calibration of automated liquid handlers and manual pipettors- Two sets of reagents- GLH-Tracker Desktop Software- MicroplatesLegal Disclaimer: For research use only, not for diagnostic procedures.
This item is not returnable. View return policy
Catalog No. NC2293256 Supplier AUTOMATION TRAINER Supplier No. ATQCKITLTAQ384
Please contact your sales representative for a quote or further ordering instructions.
The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution. Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.Characterizes accuracy and precision of volume transfersCan be used for hardware calibration of automated liquid handlers and manual pipettors- Two sets of reagents- GLH-Tracker Desktop Software- MicroplatesLegal Disclaimer: For research use only, not for diagnostic procedures.