Learn More
AUTOMATION TRAINER GLH QC Kit - Reagents Set - DMSO
SDP
Catalog No. NC3727926
Encompass_Preferred
"Quality Simplified: Automation Trainer’s GLH QC Kit allows users to characterize the error limits of volume transfers and define calibration recommendations for automated liquid handling platforms, manual pipettors, and microplate readers.The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution.Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.- Single-Dye Absorbance-Based Method- Characterizes error limits of volume transfers (1 nL to 10 mL)- Compatible with any manual or automated liquid handling instrument (up to 1536-channels)- Test the photometric accuracy, precision, and linearity of most readers with the GLH Validation Reference Plate- Low Diluent-Dispense sensitivity with GLH Verification Microplates
Catalog No. NC3727926 Supplier AUTOMATION TRAINER Supplier No. ATQCKITRSDMSO
"Quality Simplified: Automation Trainer’s GLH QC Kit allows users to characterize the error limits of volume transfers and define calibration recommendations for automated liquid handling platforms, manual pipettors, and microplate readers.The vision behind the GLH QC Kit was to provide users with a simplified, accurate, and cost-effective quality control solution.Test your automated liquid handlers, multi-channel pipettes, readers, labware, and more. Develop unique liquid classes and Optimize Assays.- Single-Dye Absorbance-Based Method- Characterizes error limits of volume transfers (1 nL to 10 mL)- Compatible with any manual or automated liquid handling instrument (up to 1536-channels)- Test the photometric accuracy, precision, and linearity of most readers with the GLH Validation Reference Plate- Low Diluent-Dispense sensitivity with GLH Verification Microplates