Learn More
Hach Company GLI pH Encap Sens-LCP-5 Wire
Catalog No. 5000207581 Shop All Hach Company Products
Encompass
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Antimony Electrode-Ceramic/Ceramic Saltbridge-10 ft cableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode
Catalog No. 50-002-07581 Supplier Hach Company Supplier No. 6028P1
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Antimony Electrode-Ceramic/Ceramic Saltbridge-10 ft cableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Antimony pH Electrode