Learn More
Hach Company GLI pH Encap Sens-LCP-5 Wire
Catalog No. 5000208091 Shop All Hach Company Products
Encompass
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Amphenol Termination-12 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-08091 Supplier Hach Company Supplier No. 6028P012
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Convertible-Amphenol Termination-12 ft CableLCP (Liquid Crystal Polymer) Convertible Mounting Style 3 5m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode