Learn More
Hach Company GLI pH Encapsulated Sensor
Catalog No. 5000207824 Shop All Hach Company Products
Encompass
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-Low pH Glass-10 ft Cable-LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode
Catalog No. 50-002-07824 Supplier Hach Company Supplier No. 6058P6
GLI pH Encapsulated Sensor-LCP Body-5 Wire-Flow-Thru Mount-Low pH Glass-10 ft Cable-LCP (Liquid Crystal Polymer) Union Mounting Style 3m Analog Cable 5-Wire with Built-In Preamplifier Low pH Glass Electrode