Learn More
Hach Company GLI-SMALL BORE ELECT. SNS
Catalog No. 5000207647 Shop All Hach Company Products
Encompass
Analog Electrodeless Conductivity Sensor-Convertible Style-PVDF Body Material-Small Bore Electrode SNS-50 ft CablePVDF Convertible Mounting Style 15m (50 ft) Analog Cable Stainless Steel Tag Electrodeless Conductivity Sensor
Catalog No. 50-002-07647 Supplier Hach Company Supplier No. 3726E2T50N
Analog Electrodeless Conductivity Sensor-Convertible Style-PVDF Body Material-Small Bore Electrode SNS-50 ft CablePVDF Convertible Mounting Style 15m (50 ft) Analog Cable Stainless Steel Tag Electrodeless Conductivity Sensor