Learn More
Aviva Systems Biology Corp GLI1 Antibody - C-terminal region (ARP32368_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32368_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GLI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GLI1. Alternate names: GLIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54547